Calpain Inhibitor III, 95+%
Calpain Inhibitor III, 95+%
Thermo Scientific Chemicals

Calpain Inhibitor III, 95+%

 
Have Questions?
Quantity:
25 mg
Catalog number J62919.MB
also known as J62919-MB
Price (USD)/ Each
145.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
145.00
Add to cart
Calpain Inhibitor III, 95+%
Catalog numberJ62919.MB
Price (USD)/ Each
145.00
-
Add to cart
Chemical Identifiers
CAS88191-84-8
IUPAC Namebenzyl N-[(1S)-2-methyl-1-{[(2S)-1-oxo-3-phenylpropan-2-yl]carbamoyl}propyl]carbamate
Molecular FormulaC22H26N2O4
InChI KeyNGBKFLTYGSREKK-PMACEKPBSA-N
SMILESCC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)N[C@@H](CC1=CC=CC=C1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
LCMSConforms to structure
Appearance (Color)White
Assay from Supplier's CofA≥95.0%
FormLyophilized solid
Inhibits calpain proteolysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C