Protamine sulfate, Thermo Scientific Chemicals
This product is animal origin and can only be shipped within the United States
Protamine sulfate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Protamine sulfate, Thermo Scientific Chemicals

Reverses the anticoagulant effects of heparin
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62926.09
also known as J62926-09
Price (USD)/ Each
182.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
215.00 
Save 32.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
182.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
215.00 
Save 32.35 (15%)
Add to cart
Protamine sulfate, Thermo Scientific Chemicals
Catalog numberJ62926.09
Price (USD)/ Each
182.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
215.00 
Save 32.35 (15%)
-
Add to cart
Chemical Identifiers
CAS9009-65-8
SMILES*
Molecular Weight (g/mol)0.00
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying5.0% max.
FormPowder
Appearance (Color)White to off-white
Assay (unspecified)Each mg neutralizes not less than 100 USP Heparin units (dry basis)
It is suitable for personal care, cosmetic and pharmaceutical applications. It is used widely to reverse the anticoagulant effects of heparin. It acts as a heparin antagonist. It is also a weak anticoagulant. It may be used for the removal of DNA from protein samples or the purification of DNA binding proteins. Protamine sulfate may also be used as an alternative to polybrene in retroviral mediated gene transfer. Protamine sulfate has antibacterial properties and inhibits pepsin activity. Protamine sulfate and other polycations have been shown to inhibit the classical pathway of complement. It neutralizes the anticoagulant activity of heparin, and inhibits turnover of lipoproteins by lipoprotein lipase. The aggregation of erythrocytes in the presence of protamine sulfate, for application to the prediction of MN phenotypes, has been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is sparingly soluble in water.

Notes
Hygroscopic. Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Kenneth Cornetta.; W.French Anderson. Protamine sulfate as an effective alternative to polybrene in retroviral-mediated gene-transfer: implications for human gene therapy. Journal of Virological Methods. 1989, 23, (2), 187-194.
  2. Sorgi, FL.; Bhattacharya, S.; Huang, L. Protamine sulfate enhances lipid-mediated gene transfer. Gene therapy . 1997, 4, (9), 961-8.