Thermo Scientific Chemicals

Anisomycin, 97%, Thermo Scientific Chemicals

Catalog number: J62964.MF
50 mg, Each
Thermo Scientific Chemicals

Anisomycin, 97%, Thermo Scientific Chemicals

Catalog number: J62964.MF
50 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62964.MF
also known as J62964-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22862-76-6
IUPAC Name
(2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
Molecular Formula
C14H19NO4
InChI Key
YKJYKKNCCRKFSL-BFHYXJOUSA-N
SMILES
COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1
Form
Powder
Assay from Supplier's CofA
≥96.0%
Appearance (Color)
White

Description

A protein synthesis inhibitor. Causes apoptosis of PC12 Cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A protein synthesis inhibitor
  • Causes apoptosis of PC12 Cells
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text