Thermo Scientific Chemicals

RBC lysis buffer for human

Catalog number: J62990.AK
250mL, Each
Thermo Scientific Chemicals

RBC lysis buffer for human

Catalog number: J62990.AK
250mL, Each
Quantity
Catalog number: J62990.AK
also known as J62990-AK
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
pH
7.0-7.6
Appearance (Color)
Clear colorless
Comment
Filtered through 0.22 micron filter.

Description

For lysis of human red blood cells and can be used to lyse reticulocytes, erythrocytes, and mature RBCs.

RBC Lysis Buffer for human is used in the isolation of deoxyribonucleic acid (DNA) and ribonucleic acid (RNA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text