Thermo Scientific Chemicals

1-Naphthyl phosphate monosodium salt monohydrate, 97+%, Thermo Scientific Chemicals

Catalog number: J63004.14
25 g, Each
Thermo Scientific Chemicals

1-Naphthyl phosphate monosodium salt monohydrate, 97+%, Thermo Scientific Chemicals

Catalog number: J63004.14
25 g, Each
Quantity
Catalog number: J63004.14
also known as J63004-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81012-89-7
Solution Test
Solubility (0.5% in water): Clear solution
Form
Solid
Assay (Titration)
≥ 97.0 %
pH
3.5 to 4.5 (5% in water)
Appearance (Color)
Off white to light pink

Description

Substrate for determination of phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text