DL-Adrenaline, Thermo Scientific Chemicals
DL-Adrenaline, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Adrenaline, Thermo Scientific Chemicals

A hormone and neurotransmitter
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number J63005.14
also known as J63005-14
Price (USD)/ Each
322.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
322.00
Add to cart
DL-Adrenaline, Thermo Scientific Chemicals
Catalog numberJ63005.14
Price (USD)/ Each
322.00
-
Add to cart
Specifications
Chemical Name or MaterialDL-Adrenaline
CAS329-65-7
Health Hazard 1H311-H315-H319-H335
Health Hazard 2GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c
View more
A hormone and neurotransmitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only