Citrate, 0.5M buffer soln., pH 2.5
Citrate, 0.5M buffer soln., pH 2.5
Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 2.5

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J63008.AK
also known as J63008-AK
Price (USD)/ Each
94.80
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
94.80
Add to cart
Citrate, 0.5M buffer soln., pH 2.5
Catalog numberJ63008.AK
Price (USD)/ Each
94.80
-
Add to cart
Specifications
ColorColorless
Concentration0.5M buffer soln.
Molecular Weight (g/mol)294.10
pH2.5
Quantity250 mL
View more
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

Solubility
It is soluble in water (29.4 g/L) at 20°C. Insoluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. YP Lee.; T Takahashi. An improved colorimetric determination of amino acids with the use of ninhydrin. Analytical Biochemistry. 1972, 14,(1), 71-77.
  2. XZ Shu.; KJ Zhu.; W Song. Novel pH-sensitive citrate cross-linked chitosan film for drug controlled release. International Journal of Pharmaceutics. 2001, 212,(1), 19-28.
Contents & Storage
Keep cold