Thermo Scientific Chemicals

Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: J63013.03
1 g, Each
Thermo Scientific Chemicals

Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: J63013.03
1 g, Each
Quantity
Catalog number: J63013.03
also known as J63013-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5541-93-5
IUPAC Name
trisodium 2-(phosphonatooxy)prop-2-enoate
Molecular Formula
C3H2Na3O6P
InChI Key
RJUFBDMHZGRGMA-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[O-]C(=O)C(=C)OP([O-])([O-])=O
Appearance (Color)
White
Assay from Supplier's CofA
≥97.0%
Form
Powder or crystals or crystalline powder
Water Content
33.0-37.0%
Infrared spectrum
Conforms

Description

When coupled with pyruvate kinase it completes an ATP-regenerating system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text