Thermo Scientific Chemicals

Arsenazo III, Thermo Scientific Chemicals

Catalog number: J63031.06
5 g, Each
Thermo Scientific Chemicals

Arsenazo III, Thermo Scientific Chemicals

Catalog number: J63031.06
5 g, Each
Quantity
Catalog number: J63031.06
also known as J63031-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1668-00-4
IUPAC Name
3,6-bis[2-(2-arsonophenyl)hydrazin-1-ylidene]-4,5-dioxo-3,4,5,6-tetrahydronaphthalene-2,7-disulfonic acid
Molecular Formula
C22H18As2N4O14S2
InChI Key
TVMZRHVOFZTNET-UHFFFAOYSA-N
SMILES
O[As](O)(=O)C1=CC=CC=C1NN=C1C(=O)C2=C(C=C1S(O)(=O)=O)C=C(C(=NNC1=CC=CC=C1[As](O)(O)=O)C2=O)S(O)(=O)=O
Appearance (Color)
Dark green to dark brown to dark red to black
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥70%
Loss on Drying
≤12.0% (200°C/constant weight)
lambda-max (UV-Vis)
528-545nm

Description

Suitable for determination of micromolar amounts of calcium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Arsenazo III is highly specific for calcium and produces absorbance peaks in the 600-660 nm range
• It reacts in slightly acidic conditions, and is soluble in alcohol and semi-soluble in water

Application

• Arsenazo III can detect calcium in permeabilized cells; it is also capable of detecting uranium and thorium
• Arsenazo III has been used in vitro to measure intracellular Ca2+ levels in live neurons alongside patch clamp methods

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text