Thermo Scientific Chemicals

Penicillin G sodium salt, Thermo Scientific Chemicals

Catalog number: J63032.22
100 g, Each
Thermo Scientific Chemicals

Penicillin G sodium salt, Thermo Scientific Chemicals

Catalog number: J63032.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63032.22
also known as J63032-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-57-8
IUPAC Name
sodium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Formula
C16H17N2NaO4S
InChI Key
FCPVYOBCFFNJFS-LQDWTQKMSA-M
SMILES
[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Appearance (Color)
White to pale cream
Form
Powder
Assay (unspecified)
Potency: ≥1500 µ/mg
pH
5.0-7.5, 5% in water, 25°C

Description

Penicillin G sodium salt is used as a β-lactam antibiotic. Penicillin G can be used as a selective agent in several types of isolation media. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
• Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis
Applications
• Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
• It can also be used to prevent infection in research animals
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text