Transfer or electro blotting buffer (10X), pH 8.5
Transfer or electro blotting buffer (10X), pH 8.5
Thermo Scientific Chemicals
Transfer or electro blotting buffer (10X), pH 8.5
Used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1L
2L
4L
Catalog number J63037.K2
also known as J63037-K2
Price (USD)/ Each
56.20
-
Add to cart
Quantity:
1L
Request bulk or custom format
Price (USD)/ Each
56.20
Add to cart
Transfer or electro blotting buffer (10X), pH 8.5
Catalog numberJ63037.K2
Price (USD)/ Each
56.20
-
Add to cart
Specifications
FormLiquid
Quantity1L
Storage RequirementsAmbient temperatures
Unit SizeEach
Transfer or electro blotting buffer (10X), pH 8.5 is used to allows protein transfer from gels to nitrocellulose or PVDF at relatively higher voltages.