General Description
• Aprotinin is a 58-amino acid polypeptide chain found in lung tissue
• Aprotinin can suppress the systemic inflammatory response by reversibly inhibiting various esterases and proteases; it acts as a broad spectrum serine protease inhibitor
Application
• Aprotinin can be used as an enzyme inhibitor in laboratory studies on chymotrypsin, kallikrein, plasmin, and others
• Aprotinin is useful for the laboratory study of post-operative bleeding and wound healing
Literature References
Parvatikar A, Vacaliuc GS, Ramanathan A, Chennubhotla SC. ANCA: Anharmonic Conformational Analysis of Biomolecular Simulations. Biophys J. 2018 May 8;114(9):2040-2043. doi: 10.1016/j.bpj.2018.03.021. PMID: 29742397; PMCID: PMC5961460.
Deloge E, Amour J, Provenchère S, Rozec B, Scherrer B, Ouattara A. Aprotinin vs. tranexamic acid in isolated coronary artery bypass surgery: A multicentre observational study. Eur J Anaesthesiol. 2017 May;34(5):280-287. doi: 10.1097/EJA.0000000000000604. PMID: 28221207.