Thermo Scientific Chemicals

L-Isoleucine, Cell Culture Reagent

Catalog number: J63045.22
100 g, Each
Thermo Scientific Chemicals

L-Isoleucine, Cell Culture Reagent

Catalog number: J63045.22
100 g, Each
Quantity
Catalog number: J63045.22
also known as J63045-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73-32-5
IUPAC Name
(2S,3S)-2-amino-3-methylpentanoic acid
Molecular Formula
C6H13NO2
InChI Key
AGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILES
CC[C@H](C)[C@H](N)C(O)=O
Heavy Metals
≤15 ppm
Impurity content
Arsenic (As): ≤1.5 ppm
Impurity content
Iron (Fe): ≤30 ppm
Optical Rotation
+38.9 to +41.8
Assay from Supplier's CofA
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text