Capsazepine, 98+%, Thermo Scientific Chemicals
Capsazepine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Capsazepine, 98+%, Thermo Scientific Chemicals

A competitive antagonist for capsaicin and resiniferatoxin
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63055.MA
also known as J63055-MA
Price (USD)/ Each
288.65
Online exclusive
321.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
288.65
Online exclusive
321.00 
Save 32.35 (10%)
Add to cart
Capsazepine, 98+%, Thermo Scientific Chemicals
Catalog numberJ63055.MA
Price (USD)/ Each
288.65
Online exclusive
321.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
CAS138977-28-3
IUPAC NameN-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-2,3,4,5-tetrahydro-1H-2-benzazepine-2-carbothioamide
Molecular FormulaC19H21ClN2O2S
InChI KeyDRCMAZOSEIMCHM-UHFFFAOYSA-N
SMILESOC1=CC2=C(CN(CCC2)C(=S)NCCC2=CC=C(Cl)C=C2)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Powder
Assay (HPLC)≥ 98.0%
FormPale cream to cream
Proton NMRConforms to structure
Solution TestSoluble in DMSO at 100mM
Capsazepine is shown to inhibit the development of mechanical hyperalgesia induced by intraplanar capsaicin injection. Capsazepine is described to block Resiniferatoxin (sc-24015)- and Capsaicin-induced contractions of guinea pig treacheal smooth muscle. Studies investigating the function of the VR1 (TRPV1 receptor) have employed Capsazepine as a tool for perturbing systemic neuronal response to Capsaicin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Capsazepine is shown to inhibit the development of mechanical hyperalgesia induced by intraplanar capsaicin injection. Capsazepine is described to block Resiniferatoxin (sc-24015)- and Capsaicin-induced contractions of guinea pig treacheal smooth muscle. Studies investigating the function of the VR1 (TRPV1 receptor) have employed Capsazepine as a tool for perturbing systemic neuronal response to Capsaicin.

Solubility
Soluble in ethanol or DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Dickenson and Dray. Selective antagonism of capsaicin by capsazepine: evidence for a spinal receptor site in capsaicin-induced antinociception. Br.J.Pharmacol. 1991, 104, (4), 1045-9.
  2. Kwak et al. A capsaicin-receptor antagonist, capsazepine, reduces inflammation-induced hyperalgesic responses in the rat: evidence for an endogenous capsaicin-like substance. Neuroscience. 1998, 86, (2), 619-26.