Thermo Scientific Chemicals

L-Cystine disodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: J63071.30
250 g, Each
Thermo Scientific Chemicals

L-Cystine disodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: J63071.30
250 g, Each
Quantity
Catalog number: J63071.30
also known as J63071-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
199329-53-8
IUPAC Name
disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate hydrate
Molecular Formula
C6H12N2Na2O5S2
InChI Key
TVPDKJXPEMKMDM-UHFFFAOYNA-L
SMILES
O.[Na+].[Na+].NC(CSSCC(N)C([O-])=O)C([O-])=O
Solution Test
Clear and complete (1.0% solution in Water)
Appearance (Color)
White to pale yellow
Form
Powder
Assay from Supplier's CofA
≥98.0% (Titration)
Water Content (Karl Fischer Titration)
≤8.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text