Thermo Scientific Chemicals

Rifamycin SV sodium, 98+%, Thermo Scientific Chemicals

Catalog number: J63095.03
1 g, Each
Thermo Scientific Chemicals

Rifamycin SV sodium, 98+%, Thermo Scientific Chemicals

Catalog number: J63095.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63095.03
also known as J63095-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14897-39-3
Assay (unspecified)
Related substances, Rifamycin S: ≤ 2.0%
Appearance (Color)
Red
Assay (unspecified)
Endotoxin: ≤ 0.50 EU/mg
Assay from Supplier's CofA
≥98%
pH
6.5 to 8.0

Description

Antibiotic effective against mycobacteria. Can also inhibit DNA polymeraseRifamycin SV sodium is an antimicrobial agent used to inhibit the DNA polymerase and RNA polymerase in biochemistry. It is used for the treatment of HIV-related Tuberculosis, traveler’s diarrhea (TD), clostridium difficile-associated disease (CDAD) and rheumatoid knee synovitis. It is utilized to study potential genes like rif15 and rif16 during the conversion of rifamycin SV into rifamycin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol and dimethyl sulfoxide.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text