Ornidazole, 99%, Thermo Scientific Chemicals
Ornidazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ornidazole, 99%, Thermo Scientific Chemicals

Interferes with electron transport in metabolic pathways
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63100.14
also known as J63100-14
Price (USD)/ Each
261.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
261.00
Add to cart
Ornidazole, 99%, Thermo Scientific Chemicals
Catalog numberJ63100.14
Price (USD)/ Each
261.00
-
Add to cart
Specifications
Chemical Name or MaterialOrnidazole
CAS16773-42-5
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
Ornidazole is a drug that cures some protozoan infections. It has been investigated for use in Crohn's disease after bowel resection. It is indicated in treatment of mixed amoebiasis, mixed amoebic dysentery, mixed giardiasis, trichomoniasis, bacterial vaginosis, Sexually transmitted diseases, infections of gynaecology, lower respiratory tract infections, ENT, surgical and dental infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. P Rutgeerts.; G Van Assche .; S Vermeire.; G D'Haens. Ornidazole for prophylaxis of postoperative Crohn's disease recurrence: a randomized, double-blind, placebo-controlled trial. Gastroenterology. 2005128 (4), 856-861.
  2. DE Schwartz.; F Jeun. Comparative pharmacokinetic studies of ornidazole and metronidazole in man. Chemotherapy. 197622 (1), 19-29 .