Thermo Scientific Chemicals

Demeclocycline hydrochloride, Thermo Scientific Chemicals

Catalog number: J63102.MD
250 mg, Each
Thermo Scientific Chemicals

Demeclocycline hydrochloride, Thermo Scientific Chemicals

Catalog number: J63102.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63102.MD
also known as J63102-MD
Price (USD)
Quantity
-
Have Questions?

Description

Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text