Calcium chloride, 1M aq. soln., Thermo Scientific Chemicals
Calcium chloride, 1M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium chloride, 1M aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Catalog number J63122.AE
also known as J63122-AE
Price (USD)/ Each
79.65
Online exclusive
88.30 
Save 8.65 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.30 
Save 8.65 (10%)
Add to cart
Calcium chloride, 1M aq. soln., Thermo Scientific Chemicals
Catalog numberJ63122.AE
Price (USD)/ Each
79.65
Online exclusive
88.30 
Save 8.65 (10%)
-
Add to cart
Chemical Identifiers
CAS10043-52-4
IUPAC Namecalcium dichloride
Molecular FormulaCaCl2
InChI KeyUXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.9-1.1M
Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong acids, borane/boron oxides, zinc, calcium oxide, methyl vinyl ether. Calcium chloride is attacked by bromine trifluoride. Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant.
RUO – Research Use Only

General References:

  1. AJ Ellis.; RM Golding. The solubility of carbon dioxide above 100 °C in water and in sodium chloride solutions. American Journal of Science. 1963261 (1), 47-60.
  2. NA Talvitie. Radiochemical determination of plutonium in environmental and biological samples by ion exchange. Analytical chemistry. 197143 (13), 1827-1830.