Thermo Scientific Chemicals

Bradykinin

Catalog number: J63131.MA
10 mg, Each
Thermo Scientific Chemicals

Bradykinin

Catalog number: J63131.MA
10 mg, Each
Quantity
Catalog number: J63131.MA
also known as J63131-MA
Price (USD)
Quantity
-
Have Questions?
Purity
≥94.0%
Appearance (Color)
White
Molecular Weight
Consistent with theoretical MW
Form
Lyophilized Powder

Description

Bradykinin is used to lower blood pressure, increase bradykinin (by inhibiting its degradation) further lowering blood pressure. Bradykinin dilates blood vessels via the release of prostacyclin, nitric oxide, and endothelium-derived hyperpolarizing factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text