Thermo Scientific Chemicals

Cyproconazole, Thermo Scientific Chemicals

Catalog number: J63142.06
5 g, Each
Thermo Scientific Chemicals

Cyproconazole, Thermo Scientific Chemicals

Catalog number: J63142.06
5 g, Each
Quantity
Catalog number: J63142.06
also known as J63142-06
Price (USD)
Quantity
-
Have Questions?

Description

Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.

Solubility
Soluble in acetone, ethanol, xylene, and dimethyl sulfoxide.

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text