2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals
2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals

A useful biological buffer
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number J63144.14
also known as J63144-14
Price (USD)/ Each
24.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
24.90
Add to cart
2-Amino-2-methyl-1,3-propanediol, 99+%, Thermo Scientific Chemicals
Catalog numberJ63144.14
Price (USD)/ Each
24.90
-
Add to cart
Chemical Identifiers
CAS115-69-5
IUPAC Name2-amino-2-methylpropane-1,3-diol
Molecular FormulaC4H11NO2
InChI KeyUXFQFBNBSPQBJW-UHFFFAOYSA-N
SMILESCC(N)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>99.0%
A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

Solubility
Soluble in water and other organic solvents.

Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, aluminum, copper, brass and strong acids.
RUO – Research Use Only

General References:

  1. Devi, T. S. R.; Kumar, J. S.; Ramkumaar, G. R. DFT simulations and vibrational spectra of 2-amino-2-methyl-1,3-propanediol. Spectrochim. Acta, Part A 2014, 133, 214-222.
  2. Mekala, P.; Kamisetty, V.; Chien, W. M.; Shi, R.; Chandra, D.; Sangwai, J.; Talekar, A.; Mishra, A. Thermodynamic modeling of binary phase diagram of 2-amino-2-methyl-1,3-propanediol and TRIS(hydroxymethyl)aminomethane system with experimental verification. Calphad 2015, 50, 126-133.