Thermo Scientific Chemicals

alpha-Ketoglutaric acid disodium salt dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: J63154.14
25 g, Each
Thermo Scientific Chemicals

alpha-Ketoglutaric acid disodium salt dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: J63154.14
25 g, Each
Quantity
Catalog number: J63154.14
also known as J63154-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
305-72-6
IUPAC Name
disodium 2-oxopentanedioate
Molecular Formula
C5H4Na2O5
InChI Key
YBGBJYVHJTVUSL-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥98.5%
Form
Powder
Loss on Drying
≤17.0%

Description

alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

Solubility
Soluble in water (1 M).

Notes
Hygroscopic. Store under inert gas. Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text