Thermo Scientific Chemicals

Naringin, Thermo Scientific Chemicals

Catalog number: J63166.22
100 g, Each
Thermo Scientific Chemicals

Naringin, Thermo Scientific Chemicals

Catalog number: J63166.22
100 g, Each
Quantity
Catalog number: J63166.22
also known as J63166-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10236-47-2
IUPAC Name
(2S)-7-{[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Molecular Formula
C27H32O14
InChI Key
DFPMSGMNTNDNHN-ZPHOTFPESA-N
SMILES
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
Assay from Supplier's CofA
≥95.0% (HPLC)
Form
Powder
Loss on Drying
≤6.0%
Appearance (Color)
Pale to dark cream

Description

A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activityNaringin is used to prepare naringin dihydrochalcone, which is about 300-1800 times sweeter than sugar. It is a citrus bioflavonoid reported to inhibit cytochrome P450 monooxygenase activity. It also acts as an inhibitor of cytochrome P450 enzymes. It is employed as an antioxidant, lipid lowering and anticancer agent. In addition to this, it is used in a study to assess aldosterone production by a human adrenocortical cell line and to improve cognitive deficits of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activityNaringin is used to prepare naringin dihydrochalcone, which is about 300-1800 times sweeter than sugar. It is a citrus bioflavonoid reported to inhibit cytochrome P450 monooxygenase activity. It also acts as an inhibitor of cytochrome P450 enzymes. It is employed as an antioxidant, lipid lowering and anticancer agent. In addition to this, it is used in a study to assess aldosterone production by a human adrenocortical cell line and to improve cognitive deficits of Alzheimer′s disease.

Solubility
Soluble in water, alcohol, acetone and warm acetic acid.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text