General Description
• Cephalexin hydrate is the hydrate of the cephalosporin antibiotic, cephalexin
• Cephalexin is a beta-lactam antibiotic that inhibits bacterial growth and division
• Cephalexin binds to penicillin-binding protein and inhibits cell wall synthesis
Application
• Cephalexin works best on gram-positive microbes
• Useful for immunology research, particularly on antibiotic allergies and cross-reactivity
Literature References
Salatin S, Lotfipour F, Jelvehgari M. Preparation and characterization of a novel thermosensitive and bioadhesive cephalexin nanohydrogel: a promising platform for topical antibacterial delivery. Expert Opin Drug Deliv. 2020 Jun;17(6):881-893. doi: 10.1080/17425247.2020.1764530. Epub 2020 May 22. PMID: 32441175.
Gwee A, Autmizguine J, Curtis N, Duffull SB. Twice- and Thrice-daily Cephalexin Dosing for Staphylococcus aureus Infections in Children. Pediatr Infect Dis J. 2020 Jun;39(6):519-522. doi: 10.1097/INF.0000000000002646. PMID: 32412727.