Thermo Scientific Chemicals

Cephalexin hydrate, 97+%, Thermo Scientific Chemicals

Catalog number: J63172.14
25 g, Each
Thermo Scientific Chemicals

Cephalexin hydrate, 97+%, Thermo Scientific Chemicals

Catalog number: J63172.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63172.14
also known as J63172-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1820673-23-1
Assay (unspecified)
97.0% min.

Description

Cephalexin is a cephalosporin antibiotic used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis. Cephalexin disrupts the synthesis of the peptidoglycan layer of bacterial cell walls which is responsible for cell wall structural integrity. Peptidoglycan synthesis is facilitated by transpeptidases known as penicillin-binding proteins (PBPs). PBPs bind to the D-Ala-D-Ala at the end of muropeptides (peptidoglycan precursors) to crosslink the peptidoglycan. Cephalexin antibiotics mimic the D-Ala-D-Ala site, thereby competitively inhibiting PBP crosslinking of peptidoglycan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cephalexin hydrate is the hydrate of the cephalosporin antibiotic, cephalexin

• Cephalexin is a beta-lactam antibiotic that inhibits bacterial growth and division

• Cephalexin binds to penicillin-binding protein and inhibits cell wall synthesis

Application

• Cephalexin works best on gram-positive microbes

• Useful for immunology research, particularly on antibiotic allergies and cross-reactivity

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text