Econazole nitrate, Thermo Scientific Chemicals
Econazole nitrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Econazole nitrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63173.14
also known as J63173-14
Price (USD)/ Each
310.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
365.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
310.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
365.00 
Save 54.35 (15%)
Add to cart
Econazole nitrate, Thermo Scientific Chemicals
Catalog numberJ63173.14
Price (USD)/ Each
310.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
365.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
CAS24169-02-6
IUPAC Name1-{2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole; nitric acid
Molecular FormulaC18H16Cl3N3O4
InChI KeyDDXORDQKGIZAME-UHFFFAOYNA-N
SMILESO[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Mass SpecConforms
FormPowder
Purity≥95.0%
Appearance (Color)White to off-white
Econazole nitrate is an antifungal medication often used as a cream to treat skin infections like tinea, ringworm and athlete's foot, personal care, cosmetic and pharmaceutical applications. It is used for studies on processes such as platelet serotonin uptake, prostanoid biosynthesis, EDHF-mediated relaxation, and Ca2+ transport pathways in thymic lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only