Thermo Scientific Chemicals
DNA Marker, high range, 1,000 Base Pair Ladder
Catalog number: J63178.S0
2 mL, Each
Thermo Scientific Chemicals
DNA Marker, high range, 1,000 Base Pair Ladder
Catalog number: J63178.S0
2 mL, Each
Quantity
Catalog number: J63178.S0
also known as J63178-S0
Price (USD)
Have Questions?
Description
DNA ladders are used in gel electrophoresis to determine the size and quantity of testing DNA fragments of genomic, plasmid, and PCR DNA. It is commonly used in the studies of plasmid DNA and PCR products. 1 kb DNA Ladder has been used as a molecular marker to determine the molecular weight and size of double stranded DNA during electrophoresis. 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb. In NA electrophoresis, five ?l of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide, SYBR™ Green, and Nancy-520.