Thermo Scientific Chemicals

Yeast Lytic Enzyme, Arthrobacter luteus

Catalog number: J63195.ME
500 mg, Each
Thermo Scientific Chemicals

Yeast Lytic Enzyme, Arthrobacter luteus

Catalog number: J63195.ME
500 mg, Each
Quantity
Catalog number: J63195.ME
also known as J63195-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37340-57-1
DNase
Trace level
Unit definition
One unit of lytic acitvity is defined as amount which indicates 30% of decrease in absorbance at A800nm
Form
Lyophilized powder
RNase
Trace level
units/mgdw
Report value

Description

Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text