Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, standard sensitivity, Thermo Scientific Chemicals

Catalog number: J63202.AP
500 mL, Each
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, standard sensitivity, Thermo Scientific Chemicals

Catalog number: J63202.AP
500 mL, Each
Quantity
Catalog number: J63202.AP
also known as J63202-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54827-17-7
IUPAC Name
3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Formula
C16H20N2
InChI Key
UAIUNKRWKOVEES-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
Form
Clear liquid
Clarity
Transparent
Composition
TMB in a dilute organic solvent with buffer, hydrogen peroxide is added at a concentration of 0.03%.
Comment
The reagent has been specifically formulated for measuring peroxidase in ELISA systems. Reaction pH is approximately pH 6.0 (Range 5.0 - 7.0)
pH
6 ± 0.5

Description

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text