Lactalbumin hydrolysate is used as a highly soluble source of amino acids and peptides. It is especially useful in fermentations, laboratory media and in tissue culture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lactalbumin hydrolysate is used as a highly soluble source of amino acids and peptides. It is especially useful in fermentations, laboratory media and in tissue culture.
Solubility
Soluble in water (130 mg/ml at 25°C).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only