Thermo Scientific Chemicals

Piroxicam, Thermo Scientific Chemicals

Catalog number: J63239.09
10 g, Each
Thermo Scientific Chemicals

Piroxicam, Thermo Scientific Chemicals

Catalog number: J63239.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63239.09
also known as J63239-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36322-90-4
IUPAC Name
(3E)-3-{hydroxy[(pyridin-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
Molecular Formula
C15H13N3O4S
InChI Key
LDJVNWFKNMYYNT-FYWRMAATSA-N
SMILES
CN1C(=C(O)NC2=CC=CC=N2)C(=O)C2=CC=CC=C2S1(=O)=O
Form
Solid
Melting Point
196-204°C
Appearance (Color)
White to pale yellow to pale green
Assay (unspecified)
≥97.0%

Description

Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.

Solubility
Soluble in water, ethanol, chloroform, ethyl acetate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text