Thermo Scientific Chemicals

4',5,7-Trihydroxyisoflavone, 99+%, Thermo Scientific Chemicals

Catalog number: J63241.03
1 g, Each
Thermo Scientific Chemicals

4',5,7-Trihydroxyisoflavone, 99+%, Thermo Scientific Chemicals

Catalog number: J63241.03
1 g, Each
Quantity
Catalog number: J63241.03
also known as J63241-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
446-72-0
IUPAC Name
5,7-dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular Formula
C15H10O5
InChI Key
TZBJGXHYKVUXJN-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
Appearance (Color)
Off-white to pale brown
Assay (HPLC)
>99.0%
Form
Powder

Description

4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.

Solubility
Soluble in chloroform, methanol. Insoluble in cold water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text