MG 132
Thermo Scientific Chemicals

MG 132

Inhibitor of calpain proteases
Have Questions?
Change viewbuttonViewtableView
Quantity:
5mg
1mg
10mg
Catalog number J63250.LB0
also known as J63250-#0
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5mg
Request bulk or custom format
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
Add to cart
MG 132
Catalog numberJ63250.LB0
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS133407-82-6
IUPAC Namebenzyl N-{3-methyl-1-[(3-methyl-1-{[(2S)-4-methyl-1-oxopentan-2-yl]carbamoyl}butyl)carbamoyl]butyl}carbamate
Molecular FormulaC26H41N3O5
InChI KeyTZYWCYJVHRLUCT-UVKLAMSESA-N
SMILESCC(C)C[C@H](NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Mass spec: Conforms
FormCrystalline solid
Thin Layer ChromatographyPurity: ≥ 98%
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

Solubility
Soluble in ethanol, chloroform, methanol, DMSO.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
Faneng Sun.; Vellareddy Anantharam.; Danhui Zhang.; Calivarathan Latchoumycandane.; Arthi Kanthasamy.; Anumantha G. Kanthasamy. Proteasome inhibitor MG-132 induces dopaminergic degeneration in cell culture and animal models.NeuroToxicology.2006, 27, (5), 807-815.
Contents & Storage
Store at -20°C