Thermo Scientific Chemicals

MG 132

Catalog number: J63250.MCR
1mg, Each
Thermo Scientific Chemicals

MG 132

Catalog number: J63250.MCR
1mg, Each
Quantity
Catalog number: J63250.MCR
also known as J63250-M^
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
133407-82-6
IUPAC Name
benzyl N-{3-methyl-1-[(3-methyl-1-{[(2S)-4-methyl-1-oxopentan-2-yl]carbamoyl}butyl)carbamoyl]butyl}carbamate
Molecular Formula
C26H41N3O5
InChI Key
TZYWCYJVHRLUCT-UVKLAMSESA-N
SMILES
CC(C)C[C@H](NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
Assay (unspecified)
Mass spec: Conforms
Form
Crystalline solid
Thin Layer Chromatography
Purity: ≥ 98%

Description

MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

Solubility
Soluble in ethanol, chloroform, methanol, DMSO.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text