Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.
Solubility
Soluble in DMSO
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Julia Penso.; Rivka Beitner. Clotrimazole and bifonazole detach hexokinase from mitochondria of melanoma cells. European Journal of Pharmacology. 1998, 342, (1), 113-117.
Maurizio Botta.; Federico Corelli.; Francesco Gasparrini.; Flavia Messina.; Claudia Mugnaini. Chiral Azole Derivatives. 4.1 Enantiomers of Bifonazole and Related Antifungal Agents: Synthesis, Configuration Assignment, and Biological Evaluation. J. Org. Chem. 2000, 65,(15), 4736-4739.