Thermo Scientific Chemicals

Bifonazole, 99%, Thermo Scientific Chemicals

Catalog number: J63253.06
5 g, Each
Thermo Scientific Chemicals

Bifonazole, 99%, Thermo Scientific Chemicals

Catalog number: J63253.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63253.06
also known as J63253-06
Price (USD)
Quantity
-
Have Questions?

Description

Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.

Solubility
Soluble in DMSO

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text