Thermo Scientific Chemicals

MES, 0.2M buffer soln., pH 5.5

Catalog number: J63257.AP
500 mL, Each
Thermo Scientific Chemicals

MES, 0.2M buffer soln., pH 5.5

Catalog number: J63257.AP
500 mL, Each
Quantity
Catalog number: J63257.AP
also known as J63257-AP
Price (USD)
Quantity
-
Have Questions?

Description

MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text