N,N'-Methylenebisacrylamide, 2% soln.
N,N'-Methylenebisacrylamide, 2% soln.
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 2% soln.

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J63265.K2
also known as J63265-K2
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
Add to cart
N,N'-Methylenebisacrylamide, 2% soln.
Catalog numberJ63265.K2
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Chemical Identifiers
CAS110-26-9
IUPAC NameN-[(prop-2-enamido)methyl]prop-2-enamide
Molecular FormulaC7H10N2O2
InChI KeyZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILESC=CC(=O)NCNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration2% w/v N,N'-methylenebisacrylamide in water
Appearance (Color)Clear, colorless
FormLiquid
CommentPrepared in 18.2 megohms water and filtered through a 0.22micron filter.
Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis. Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis
  • Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels
RUO – Research Use Only