Thermo Scientific Chemicals

Chymostatin

Catalog number: J63275.LB0
5mg, Each
Thermo Scientific Chemicals

Chymostatin

Catalog number: J63275.LB0
5mg, Each
Quantity
Catalog number: J63275.LB0
also known as J63275-#0
Price (USD)
Quantity
-
Have Questions?
Absorbance
Percentage of Component C (Isoleucine): Report
Assay from Suppliers CofA
Percentage Component A (Leucine): Report
Form
Powder
Appearance (Color)
White to off-white
Assay from Suppliers CofA
Activity: Report Value

Description

A highly specific chymotrypsin inhibitor produced by actinomycetes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text