General Description
• Fibrinogen exists in blood as a dimer of three bonded, non-identical chains, FGA, FGG, and FGB
• Fibrinogen is cleaved by thrombin to form fibrin, the most abundant component of blood clots
Application
• Fibrinogen can be used in a laboratory setting to form polymer hydrogels, particularly for use as a tissue scaffold
• Fibrinogen may be useful in the research of neurodegeneration and brain injury; some evidence shows that it has a signaling role in neural progenitor cells
General References: