General Description
• BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
• Acrylamide is a crystalline amide that forms polymer structures at room temperature
• Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation
Application
• Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
Literature References
Sunyer R, Jin AJ, Nossal R, Sackett DL. Fabrication of hydrogels with steep stiffness gradients for studying cell mechanical response. PLoS One. 2012;7(10):e46107. doi: 10.1371/journal.pone.0046107. Epub 2012 Oct 4. PMID: 23056241; PMCID: PMC3464269.
Diouf A, Bouchikhi B, El Bari N. A nonenzymatic electrochemical glucose sensor based on molecularly imprinted polymer and its application in measuring saliva glucose. Mater Sci Eng C Mater Biol Appl. 2019 May;98:1196-1209. doi: 10.1016/j.msec.2019.01.001. Epub 2019 Jan 11. PMID: 30813003.