Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals
RIPA buffer
Catalog number: J63306.AK
250 mL, Each
Thermo Scientific Chemicals
RIPA buffer
Catalog number: J63306.AK
250 mL, Each
Quantity
Catalog number: J63306.AK
also known as J63306-AK
Price (USD)
Have Questions?
Description
RIPA is used as a high quality, ready to use mammalian cell and tissue lysis buffer with protease inhibitors included, is used to lyse cells and tissue, for radio immunoprecipitation assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Sorbitan monooleate is derived from sorbitol and is a synthetic fatty acid ester that presents emulsifying, dispersing, and wetting activities
Application
• Sorbitan monooleate is used in yeast laboratory strains to protect them from excess drying and also helps rehydrate yeast cells
• It can be used as a dispersant and as a corrosion inhibitor
• Sorbitan monooleate has important uses as an excipient and intravenous osmotic diuretic