General Description
• Sorbitan monooleate is derived from sorbitol and is a synthetic fatty acid ester that presents emulsifying, dispersing, and wetting activities
Application
• Sorbitan monooleate is used in yeast laboratory strains to protect them from excess drying and also helps rehydrate yeast cells
• It can be used as a dispersant and as a corrosion inhibitor
• Sorbitan monooleate has important uses as an excipient and intravenous osmotic diuretic