Thermo Scientific Chemicals

3,3',5-Triiodo-L-thyronine sodium salt, Thermo Scientific Chemicals

Catalog number: J63312.03
1 g, Each
Thermo Scientific Chemicals

3,3',5-Triiodo-L-thyronine sodium salt, Thermo Scientific Chemicals

Catalog number: J63312.03
1 g, Each
Quantity
Catalog number: J63312.03
also known as J63312-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55-06-1
IUPAC Name
sodium (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
Molecular Formula
C15H11I3NNaO4
InChI Key
SBXXSUDPJJJJLC-YDALLXLXSA-M
SMILES
[Na+].N[C@@H](CC1=CC(I)=C(OC2=CC=C(O)C(I)=C2)C(I)=C1)C([O-])=O
Appearance (Color)
White to cream or pale yellow to pale brown
Assay from Supplier's CofA
≥97.0% (HPLC)
Water Content (Karl Fischer Titration)
≤4%
Optical Rotation
+18 to +22? [c=2, 1.2M HCl and Ethanol (1:4)]
Form
Crystals or powder or crystalline powder

Description

L-3,3',5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-3,3′,5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.

Solubility
Partly miscible in water. Soluble in dimethyl sulfoxide, methanol, sodium hydroxide and ethanol. Practically insoluble in most other organic solvents.

Notes
Store at -20°C. Incompatible with oxidizing agents and heat. Light sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text