Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 7.5

Catalog number: J63349.AK
250 mL, Each
Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 7.5

Catalog number: J63349.AK
250 mL, Each
Quantity
Catalog number: J63349.AK
also known as J63349-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7558-80-7
IUPAC Name
sodium dihydrogen phosphate
Molecular Formula
H2NaO4P
InChI Key
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES
[Na+].OP(O)([O-])=O
Form
Liquid
pH
7.5 ± 0.15 (25 °C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text