Phosphate, 0.5M buffer soln., pH 7.5
Phosphate, 0.5M buffer soln., pH 7.5
Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 7.5

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number J63349.AP
also known as J63349-AP
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
Add to cart
Phosphate, 0.5M buffer soln., pH 7.5
Catalog numberJ63349.AP
Price (USD)/ Each
98.65
Online exclusive
109.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7558-80-7
IUPAC Namesodium dihydrogen phosphate
Molecular FormulaH2NaO4P
InChI KeyAJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
pH7.5 ± 0.15 (25 °C)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures