Potassium acetate, 8M aq. soln., Thermo Scientific Chemicals
Potassium acetate, 8M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium acetate, 8M aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63372.AE
also known as J63372-AE
Price (USD)/ Each
59.70
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
59.70
Add to cart
Potassium acetate, 8M aq. soln., Thermo Scientific Chemicals
Catalog numberJ63372.AE
Price (USD)/ Each
59.70
-
Add to cart
Specifications
Chemical Name or MaterialPotassium acetate
CAS127-08-2
Recommended StorageAmbient temperatures
TSCAYes
Molecular FormulaC2H3KO2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only