Thermo Scientific Chemicals

Cytidine-5'-monophosphate disodium salt, 99+%

Catalog number: J63376.03
1 g, Each
Thermo Scientific Chemicals

Cytidine-5'-monophosphate disodium salt, 99+%

Catalog number: J63376.03
1 g, Each
Quantity
Catalog number: J63376.03
also known as J63376-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6757-06-8
IUPAC Name
disodium [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Formula
C9H12N3Na2O8P
InChI Key
INTPYBRGLGSMRA-IJRYJGAHNA-L
SMILES
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
Appearance (Color)
White
Form
Powder
Assay (HPLC)
>99.0%
Water Content (Karl Fischer Titration)
27.5% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text