Thermo Scientific Chemicals

trans-Zeatin-riboside, Thermo Scientific Chemicals

Catalog number: J63382.MB
25 mg, Each
Thermo Scientific Chemicals

trans-Zeatin-riboside, Thermo Scientific Chemicals

Catalog number: J63382.MB
25 mg, Each
Quantity
Catalog number: J63382.MB
also known as J63382-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6025-53-2
IUPAC Name
(2R,3R,4S,5R)-2-(6-{[(2E)-4-hydroxy-3-methylbut-2-en-1-yl]amino}-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Formula
C15H21N5O5
InChI Key
GOSWTRUMMSCNCW-HNNGNKQASA-N
SMILES
CC(CO)=C/CNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Loss on Drying
≤0.5%
Melting Point
178.0-182.0°C
Proton NMR
Conforms to structure
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder

Description

trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.

Solubility
Soluble in water, ethanol, and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text